Methyl trans-4-bromocinnamate 98%

Lieferant: Apollo Scientific

OR15501-250MG
APOSOR15501-250MGEA 21.9 EUR
APOSOR15501-250MG
Methyl trans-4-bromocinnamate 98%
Methyl trans-4-bromocinnamate
Formel: C₁₀H₉BrO₂
MDL: MFCD00157162
CAS-Nummer: 71205-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR