Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(1E)-2-Cyano-3-methoxy-3-oxoprop-1-en-1-yl]anisole, Methyl (2E)-2-cyano-3-(4-methoxyphenyl)prop-2-enoate, Methyl trans-α-cyano-4-methoxycinnamate

OR15502-1G
APOSOR15502-1GEA 29 EUR
APOSOR15502-1G
Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate
Methyl (E)-2-cyano-3-(4-methoxyphenyl)acrylate
Formel: C₁₂H₁₁NO₃
Molecular Weight: 217,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00630908
CAS-Nummer: 14479-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR