Methyl-3-(4-cyanophenyl)-2-oxiranecarboxylate
Lieferant: Apollo Scientific
|
Gefahr
|
OR15503-1G
APOSOR15503-1GEA
538
EUR
APOSOR15503-1G
Methyl-3-(4-cyanophenyl)-2-oxiranecarboxylate
Methyl-3-(4-cyanophenyl)-2-oxiranecarboxylate
Formel:
C₁₁H₉NO₃ Molecular Weight: 203,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864842 CAS-Nummer: 108492-59-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...