Methyl-4-(4-methoxyphenyl)-2-oxo-3-butenoate

Lieferant: Apollo Scientific
Achtung

OR15504-1G
APOSOR15504-1GEA 270 EUR
APOSOR15504-1G
Methyl-4-(4-methoxyphenyl)-2-oxo-3-butenoate
Methyl-4-(4-methoxyphenyl)-2-oxo-3-butenoate
Formel: C₁₂H₁₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09864843
CAS-Nummer: 105213-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR