4-Amino-5-bromo-2-chloropyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR15511-1G
APOSOR15511-1GEA
21.9
EUR
APOSOR15511-1G
4-Amino-5-bromo-2-chloropyrimidine
4-Amino-5-bromo-2-chloropyrimidine
Formel:
C₄H₃BrClN₃ Molecular Weight: 208,44 g/mol Schmelzpunkt: 189…191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07782040 CAS-Nummer: 205672-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...