5-Brom-2-(isopropylamino)pyrimidin

Lieferant: Apollo Scientific
Achtung

Synonyme: (5-Bromopyrimidin-2-yl)isopropylamine

OR15515-250MG
APOSOR15515-250MGEA 21.9 EUR
APOSOR15515-250MG
5-Brom-2-(isopropylamino)pyrimidin
5-Brom-2-(isopropylamino)pyrimidin
Formel: C₇H₁₀BrN₃
Molecular Weight: 216,0784 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00483159
CAS-Nummer: 77476-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR