4-Iodo-6-methoxypyrimidine

Lieferant: Apollo Scientific
Gefahr

OR15524-1G
APOSOR15524-1GEA 105 EUR
APOSOR15524-1G
4-Iodo-6-methoxypyrimidine
4-Iodo-6-methoxypyrimidine
Formel: C₅H₅IN₂O
Molecular Weight: 236,0100 g/mol
Schmelzpunkt: 68…69,5 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07787396
CAS-Nummer: 161489-05-0
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR