2-Methylthiopyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Methylsulphanyl)pyrimidine
OR15527-250MG
APOSOR15527-250MGEA
53.8
EUR
APOSOR15527-250MG
2-Methylthiopyrimidine 97%
2-Methylthiopyrimidine
Formel:
C₅H₆N₂S Molecular Weight: 126,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025802 CAS-Nummer: 823-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...