2-(Methylthio)-5-pyrimidinboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-(Methylthio)pyrimidin-5-yl)boronic acid, 2-(Methylthio)pyrimidine-5-boronic acid
OR15528-250MG
APOSOR15528-250MGEA
21.9
EUR
APOSOR15528-250MG
2-(Methylthio)-5-pyrimidinboronsäure 97%
2-(Methylthio)-5-pyrimidineboronic acid
Formel:
C₅H₇BN₂O₂S Molecular Weight: 170,0000 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07375142 CAS-Nummer: 348098-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...