Ethyl-2,5-dibromothiazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2,5-dibromothiazole-4-carboxylate

OR15536-500MG
APOSOR15536-500MGEA 21.9 EUR
APOSOR15536-500MG
Ethyl-2,5-dibromothiazole-4-carboxylate
Ethyl-2,5-dibromothiazole-4-carboxylate
Formel: C₆H₅Br₂NO₂S
Molecular Weight: 314,9800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265487
CAS-Nummer: 208264-60-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR