2-Acetamido-5-bromthiazol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(5-bromothiazol-2-yl)acetamide
OR15548-1G
OR15548-5G
OR15548-250MG
APOSOR15548-1GEA
94.4
EUR
APOSOR15548-1G
APOSOR15548-5G
APOSOR15548-250MG
2-Acetamido-5-bromthiazol
2-Acetamido-5-bromthiazol
Formel:
C₅H₅BrN₂OS Schmelzpunkt: 198…201 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00017336 CAS-Nummer: 7336-54-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...