Methyl-2-chloro-5-isopropylthiazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-chloro-5-isopropylthiazole-4-carboxylate

OR15558-1G
APOSOR15558-1GEA 72.5 EUR
APOSOR15558-1G
Methyl-2-chloro-5-isopropylthiazole-4-carboxylate
Methyl-2-chloro-5-isopropylthiazole-4-carboxylate
Formel: C₈H₁₀ClNO₂S
Molecular Weight: 219,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265506
CAS-Nummer: 81569-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR