4-Bromo-2-(thiomethyl)thiazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-(methylthio)thiazole
OR15564-1G
APOSOR15564-1GEA
75.4
EUR
APOSOR15564-1G
4-Bromo-2-(thiomethyl)thiazole
4-Bromo-2-(thiomethyl)thiazole
Formel:
C₄H₄BrNS₂ Molecular Weight: 210.12 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD07787376 CAS-Nummer: 204513-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...