Dimethyl-2-bromo-1H-imidazole-4,5-dicarboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Dimethyl 2-bromo-1H-imidazole-4,5-dicarboxylate

OR15573-1G
APOSOR15573-1GEA 29 EUR
APOSOR15573-1G
Dimethyl-2-bromo-1H-imidazole-4,5-dicarboxylate
Dimethyl-2-bromo-1H-imidazole-4,5-dicarboxylate
Formel: C₇H₇BrN₂O₄
Molecular Weight: 263,05 g/mol
Schmelzpunkt: 99…102 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09702026
CAS-Nummer: 705280-65-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR