(4-Methyl-1H-imidazol-5-yl)methanolhydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxymethyl-5-methylimidazole hydrochloride

OR15574-25G
APOSOR15574-25GEA 21.9 EUR
APOSOR15574-25G
(4-Methyl-1H-imidazol-5-yl)methanolhydrochlorid
(4-Methyl-1H-imidazol-5-yl)methanol hydrochloride
Formel: C₅H₉ClN₂O
Molecular Weight: 148,5908 g/mol
Schmelzpunkt: 233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00040986
CAS-Nummer: 38585-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR