Methyl-2-chlorooxazole-4-carboxylate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 2-chlorooxazole-4-carboxylate

OR15582-25G OR15582-5G
APOSOR15582-25GEA 719 EUR
APOSOR15582-25G APOSOR15582-5G
Methyl-2-chlorooxazole-4-carboxylate 98%
Methyl-2-chlorooxazole-4-carboxylate
Formel: C₅H₄ClNO₃
Molecular Weight: 161.55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09265505
CAS-Nummer: 934236-35-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR