3-Bromo-5-(2-hydroxyethyl)isoxazole

Lieferant: Apollo Scientific

Synonyme: 2-(3-Bromoisoxazol-5-yl)ethanol

OR15589-1G
APOSOR15589-1GEA 317 EUR
APOSOR15589-1G
3-Bromo-5-(2-hydroxyethyl)isoxazole
3-Bromo-5-(2-hydroxyethyl)isoxazole
Formel: C₅H₆BrNO₂
Molecular Weight: 192,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025786
CAS-Nummer: 105175-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR