2-Pyridinboronsäurepinakolester 96%

Lieferant: Apollo Scientific
Achtung

OR15598-1G
APOSOR15598-1GEA 183 EUR
APOSOR15598-1G
2-Pyridinboronsäurepinakolester 96%
2-Pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₂
Lagertemperatur: Gefrierschrank
MDL: MFCD04039342
CAS-Nummer: 874186-98-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR