2-Brom-4-chlorpyridin 97%

Lieferant: Apollo Scientific

OR15630-25G
APOSOR15630-25GEA 78.3 EUR
APOSOR15630-25G
2-Brom-4-chlorpyridin 97%
2-Bromo-4-chloropyridine
Formel: C₅H₃BrClN
Lagertemperatur: Raumtemperatur
MDL: MFCD03840757
CAS-Nummer: 22918-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR