5-Brom-2-(methylthio)pyridin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Bromo-2-methylthiopyridine, 2-Methylthio-5-bromopyridine
OR15634-1G
APOSOR15634-1GEA
21.9
EUR
APOSOR15634-1G
5-Brom-2-(methylthio)pyridin 98%
5-Brom-2-(methylthio)pyridin
Formel:
C₆H₆BrNS Molecular Weight: 204,0875 g/mol Schmelzpunkt: 36…41 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411571 CAS-Nummer: 51933-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...