(R)-Piperazine-2-carboxylic acid dihydrochloride

Lieferant: Apollo Scientific
Achtung

OR15637-1G
APOSOR15637-1GEA 21.9 EUR
APOSOR15637-1G
(R)-Piperazine-2-carboxylic acid dihydrochloride
(R)-Piperazine-2-carboxylic acid dihydrochloride
Formel: C₅H₁₂Cl₂N₂O₂
Molecular Weight: 203,0700 g/mol
Schmelzpunkt: 265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03840829
CAS-Nummer: 126330-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR