(S)-Piperazine-2-carboxylic acid dihydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S)-piperazine-2-carboxylic acid dihydrochloride

OR15638-5G
APOSOR15638-5GEA 43.5 EUR
APOSOR15638-5G
(S)-Piperazine-2-carboxylic acid dihydrochloride 95%
(S)-Piperazine-2-carboxylic acid dihydrochloride
Formel: C₅H₁₂Cl₂N₂O₂
Molecular Weight: 203.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632104
CAS-Nummer: 158663-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR