(S)-(-)-1-Boc-3-methylpiperazin
Lieferant: Apollo Scientific
|
Gefahr
|
OR15643-25G
APOSOR15643-25GEA
0
EUR
APOSOR15643-25G
(S)-(-)-1-Boc-3-methylpiperazin
(S)-(-)-1-Boc-3-methylpiperazin
Formel:
C₁₀H₂₀N₂O₂ Schmelzpunkt: 38…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683204 CAS-Nummer: 147081-29-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...