(S)-1-Boc-2-isobutylpiperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-tert-Butyl 2-isobutylpiperazine-1-carboxylate, (2S)-2-Isobutylpiperazine, N1-BOC protected

OR15645-250MG
APOSOR15645-250MGEA 21.9 EUR
APOSOR15645-250MG
(S)-1-Boc-2-isobutylpiperazine
(S)-1-Boc-2-isobutylpiperazine
Formel: C₁₃H₂₆N₂O₂
Molecular Weight: 242,36 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03787916
CAS-Nummer: 674792-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR