(S)-1-Boc-2-isopropylpiperazine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-1-Boc-2-Isopropylpiperazine, (2S)-2-Isopropylpiperazine, N1-BOC protected

OR15647-100MG
APOSOR15647-100MGEA 21.9 EUR
APOSOR15647-100MG
(S)-1-Boc-2-isopropylpiperazine
(S)-1-Boc-2-isopropylpiperazine
Formel: C₁₂H₂₄N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03787918
CAS-Nummer: 674792-05-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR