(S)-3-Amino-1-benzylpiperidine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-1-Benzylpiperidin-3-amine
OR15667-250MG
APOSOR15667-250MGEA
21.9
EUR
APOSOR15667-250MG
(S)-3-Amino-1-benzylpiperidine 98%
(S)-3-Amino-1-benzylpiperidine
Formel:
C₁₂H₁₈N₂ Molecular Weight: 190,29 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093375 CAS-Nummer: 168466-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...