2-Methyl-2-propanyl-3-(aminomethyl)-1-piperidincarboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
tert-Butyl 3-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 3-(Aminomethyl)-1-piperidinecarboxylate, 3-(Aminomethyl)-1-Boc-piperidine, 3-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, 3-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester
OR15670-1G
APOSOR15670-1GEA
52.2
EUR
APOSOR15670-1G
2-Methyl-2-propanyl-3-(aminomethyl)-1-piperidincarboxylat
tert-Butyl-3-(aminomethyl)piperidine-1-carboxylate
Formel:
C₁₁H₂₂N₂O₂ Molecular Weight: 214.31 g/mol Dichte: 0.995 g/cm³ (25 °C) Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01317792 CAS-Nummer: 162167-97-7 UN: 3082 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...