(S)-(+)-1-Boc-3-hydroxypyrrolidin 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate

OR15680-100G OR15680-10G OR15680-25G
APOSOR15680-100GEA 85.7 EUR
APOSOR15680-100G APOSOR15680-10G APOSOR15680-25G
(S)-(+)-1-Boc-3-hydroxypyrrolidin 99%
(S)-(+)-1-Boc-3-hydroxypyrrolidin

Chiral intermediate for pharmaceuticals and agrochemicals.

Formel: C₉H₁₇NO₃
Schmelzpunkt: 62…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01317839
CAS-Nummer: 101469-92-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR