(S)-1-Boc-2-(aminomethyl)pyrrolidine

Lieferant: Apollo Scientific
Gefahr

Synonyme: (S)-2-(Aminomethyl)-1-Boc-pyrrolidine, (S)-2-Aminomethyl-1-Boc-pyrrolidine, tert-Butyl (S)-2-(aminomethyl)-1-pyrrolidinecarboxylate, (S)-1-Boc-2-(Aminomethyl)pyrrolidine, (2S)-2-(Aminomethyl)pyrrolidine, N1-BOC protected, (S)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (S)-2-(Aminomethyl)-1-pyrrolidinecarboxylic acid tert-Butyl Ester

OR15683-5G
APOSOR15683-5GEA 58 EUR
APOSOR15683-5G
(S)-1-Boc-2-(aminomethyl)pyrrolidine
(S)-1-Boc-2-(aminomethyl)pyrrolidine
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200.28 g/mol
Siedepunkt: 112 °C/ 0.75mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD03419257
CAS-Nummer: 119020-01-8
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR