(R)-2-Methylpyrrolidine-p-toluenesulphonate

Lieferant: Apollo Scientific
Achtung

OR15684-1G
APOSOR15684-1GEA 101 EUR
APOSOR15684-1G
(R)-2-Methylpyrrolidine-p-toluenesulphonate
(R)-2-Methylpyrrolidine-p-toluenesulphonate
Formel: C₁₂H₁₉NO₃S
Molecular Weight: 257,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368911
CAS-Nummer: 204387-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR