(2S)-2-Methylpyrrolidine toluene-4-sulphonate

Lieferant: Apollo Scientific
Achtung

Synonyme: (2S)-2-Methylpyrrolidine tosylate (2S)-2-Methylpyrrolidine 4-methylbenzenesulphonate

OR15685-1G
APOSOR15685-1GEA 159 EUR
APOSOR15685-1G
(2S)-2-Methylpyrrolidine toluene-4-sulphonate
(2S)-2-Methylpyrrolidine toluene-4-sulphonate
Formel: C₅H₁₁N·C₇H₈O₃S
Molecular Weight: 257,349 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07368912
CAS-Nummer: 1212353-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR