2-Chloro-5-(methylthio)benzoic acid 97%, tech

Lieferant: Apollo Scientific
Achtung

OR15741-5G
APOSOR15741-5GEA 0 EUR
APOSOR15741-5G
2-Chloro-5-(methylthio)benzoic acid 97%, tech
2-Chloro-5-(methylthio)benzoic acid
Formel: C₈H₇ClO₂S
Schmelzpunkt: 136…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009721
CAS-Nummer: 51546-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR