7-Azaindole-5-boronic acid pinacol ester
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Azaindole-5-boronic Acid Pinacol Ester
OR15744-5G
APOSOR15744-5GEA
95.7
EUR
APOSOR15744-5G
7-Azaindole-5-boronic acid pinacol ester
7-Azaindole-5-boronic acid pinacol ester
Formel:
C₁₃H₁₇BN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08060937 CAS-Nummer: 754214-56-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...