1-Methylpyrrole-2-carbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR15749-1G
APOSOR15749-1GEA
589
EUR
APOSOR15749-1G
1-Methylpyrrole-2-carbonyl chloride
1-Methylpyrrole-2-carbonyl chloride
Formel:
C₆H₆ClNO Molecular Weight: 143.57 g/mol Siedepunkt: 84 °C/15mmHg Lagertemperatur: Gefrierschrank |
MDL:
MFCD02094021 CAS-Nummer: 26214-68-6 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...