Ethyl 3,4-dimethyl-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR15759-1G
APOSOR15759-1GEA 0 EUR
APOSOR15759-1G
Ethyl 3,4-dimethyl-1H-pyrrole-2-carboxylate
Ethyl-3,4-dimethyl-1H-pyrrole-2-carboxylate
Formel: C₉H₁₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00030365
CAS-Nummer: 938-75-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR