1-[2-(Benzhydryloxy)ethyl]-1,4-diazepane

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-[2-(Diphenylmethoxy)ethyl]homopiperazine

OR1581-500MG
APOSOR1581-500MGEA 79.9 EUR
APOSOR1581-500MG
1-[2-(Benzhydryloxy)ethyl]-1,4-diazepane
1-[2-(Benzhydryloxy)ethyl]-1,4-diazepane
Formel: C₂₀H₂₆N₂O
Molecular Weight: 310.44 g/mol
Siedepunkt: 183 °C/0.5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00729027
CAS-Nummer: 150557-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR