4-Allyl-5-(N-tert-butoxycarbonylaminomethyl)-1,2,4-triazole-3-thiol 95+%
Lieferant: Apollo Scientific
OR1582-1G
APOSOR1582-1GEA
87
EUR
APOSOR1582-1G
4-Allyl-5-(N-tert-butoxycarbonylaminomethyl)-1,2,4-triazole-3-thiol 95+%
4-Allyl-5-(N-tert-butoxycarbonylaminomethyl)-1,2,4-triazole-3-thiol
Formel:
C₁₁H₁₈N₄O₂S Molecular Weight: 270,351 g/mol Schmelzpunkt: 150…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570690 CAS-Nummer: 306935-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...