1,3-Thiazole-4-thiocarboxamide

Lieferant: Apollo Scientific
Achtung

OR15911-1G
APOSOR15911-1GEA 120 EUR
APOSOR15911-1G
1,3-Thiazole-4-thiocarboxamide
1,3-Thiazole-4-thiocarboxamide
Formel: C₄H₄N₂S₂
Lagertemperatur: Kühlschrank
MDL: MFCD04115283
CAS-Nummer: 80653-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR