2-Methylthiazole-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR15916-1G
APOSOR15916-1GEA 36.4 EUR
APOSOR15916-1G
2-Methylthiazole-5-carbaldehyde
2-Methylthiazole-5-carbaldehyde
Formel: C₅H₅NOS
Molecular Weight: 127,1600 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09864617
CAS-Nummer: 1003-60-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR