Ethyl-3-aminoisonicotinat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-amino-4-pyridinecarboxylate, Ethyl 3-Aminoisonicotinate, 3-Aminoisonicotinic acid ethyl ester, Ethyl 3-Aminopyridine-4-carboxylate, 3-Aminopyridine-4-carboxylic acid ethyl ester

OR1592-1G
APOSOR1592-1GEA 21.9 EUR
APOSOR1592-1G
Ethyl-3-aminoisonicotinat
Ethyl-3-amino-4-pyridinecarboxylate
Formel: C₈H₁₀N₂O₂
Schmelzpunkt: 170…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03788270
CAS-Nummer: 14208-83-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR