Methyl-4-hydroxypyrimidine-5-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-hydroxypyrimidine-5-carboxylate

OR15941-1G
APOSOR15941-1GEA 116 EUR
APOSOR15941-1G
Methyl-4-hydroxypyrimidine-5-carboxylate 95%
Methyl-4-hydroxypyrimidine-5-carboxylate
Formel: C₆H₆N₂O₃
Molecular Weight: 154,1200 g/mol
Schmelzpunkt: 209…210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01632117
CAS-Nummer: 4774-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR