2-(Aminomethyl)-3-methylfuran 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: (3-Methylfur-2-yl)methylamine, 3-Methylfurfurylamine

OR15948-50MG
APOSOR15948-50MGEA 21.9 EUR
APOSOR15948-50MG
2-(Aminomethyl)-3-methylfuran 97%
2-(Aminomethyl)-3-methylfuran
Formel: C₆H₉NO
Molecular Weight: 111,142 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09811371
CAS-Nummer: 388072-09-1
UN: 2733
ADR: 3+8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR