4-[(Methylamino)methyl]tetrahydro-2H-thiopyran 97%

Lieferant: Apollo Scientific

Synonyme: N-Methyl(tetrahydro-2H-thiopyran-4-yl)methylamine

OR15951-1G
APOSOR15951-1GEA 296 EUR
APOSOR15951-1G
4-[(Methylamino)methyl]tetrahydro-2H-thiopyran 97%
4-[(Methylamino)methyl]tetrahydro-2H-thiopyran
Formel: C₇H₁₅NS
Molecular Weight: 145,266 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09879924
CAS-Nummer: 950603-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR