Tetrahydro-2H-thiopyran-4-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carboxytetrahydro-2H-thiopyran, 4-Carboxythiane, Tetrahydrothiopyran-4-carboxylic Acid

OR15954-250MG
APOSOR15954-250MGEA 191 EUR
APOSOR15954-250MG
Tetrahydro-2H-thiopyran-4-carboxylic acid 97%
Tetrahydro-2H-thiopyran-4-carboxylic acid
Formel: C₆H₁₀O₂S
Molecular Weight: 146.207 g/mol
Schmelzpunkt: 111…112.5 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09879921
CAS-Nummer: 89489-53-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR