4-Bromothiophene-3-carbonyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

OR15958-1G
APOSOR15958-1GEA 174 EUR
APOSOR15958-1G
4-Bromothiophene-3-carbonyl chloride 97%
4-Bromothiophene-3-carbonyl chloride
Formel: C₅H₂BrClOS
Molecular Weight: 225,491 g/mol
Schmelzpunkt: 67,5…69 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08060080
CAS-Nummer: 72899-51-5
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR