4-Methylphenylglyoxal hydrate 95%

Lieferant: Apollo Scientific
Achtung

OR1596-250MG
APOSOR1596-250MGEA 21.9 EUR
APOSOR1596-250MG
4-Methylphenylglyoxal hydrate 95%
4-Methylphenylglyoxal hydrate
Formel: C₉H₁₀O₃
Molecular Weight: 166.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00234532
CAS-Nummer: 16208-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR