4-Bromothien-3-yl isocyanate 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Bromo-4-isocyanatothiophene

OR15962-1G
APOSOR15962-1GEA 497 EUR
APOSOR15962-1G
4-Bromothien-3-yl isocyanate 95%
4-Bromothien-3-yl isocyanate

May form small amount of sediment

Formel: C₅H₂BrNOS
Molecular Weight: 204,044 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09879904
CAS-Nummer: 937795-88-5
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR