Methyl-2-bromofuran-3-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

OR15964-1G OR15964-250MG OR15964-100MG
APOSOR15964-1GEA 273 EUR
APOSOR15964-1G APOSOR15964-250MG APOSOR15964-100MG
Methyl-2-bromofuran-3-carboxylate 97%
Methyl-2-bromofuran-3-carboxylate
Formel: C₆H₅BrO₃
Molecular Weight: 205.01 g/mol
Schmelzpunkt: 49…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09879944
CAS-Nummer: 197846-06-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR