1-Hydroxy-1-cyclopropanecarboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR15989-1G OR15989-5G OR15989-25G
APOSOR15989-1GEA 21.9 EUR
APOSOR15989-1G APOSOR15989-5G APOSOR15989-25G
1-Hydroxy-1-cyclopropanecarboxylic acid 98%
Formel: C₄H₆O₃
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010797
CAS-Nummer: 17994-25-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR