3-(2-Chlorophenyl)-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
OR16-1G
APOSOR16-1GEA
132
EUR
APOSOR16-1G
3-(2-Chlorophenyl)-1H-pyrazole
3-(2-Chlorophenyl)-1H-pyrazole
Formel:
C₉H₇ClN₂ Schmelzpunkt: 76…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00111882 CAS-Nummer: 59843-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...